ELISA Carbonate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developingRead more
Have Questions?
Catalog Number
Quantity
CB01100
100 mL
Catalog number CB01100
Price (USD)
-
Contact Us
Quantity:
100 mL
ELISA Carbonate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA. The buffer is a 50 mM carbonate buffer, pH 9.4.