Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | PEEK |
Range | 0 to 5000 µS/cm; 0 to 18.2 MW·cm |
Electrode | Two electrode measurement with 0.01, 0.1, and 1.0 cell constants |
Max. Pressure | 150psig (at 75°C); 150psig (at 95°C) |
Type | Conductivity Resistivity Sensors, PEEK |
For Use With (Equipment) | AquaSensors™ DataStick™ Analytical Measurement System |
Flow Rate | 10 ft./sec. |
Resolution | 4.5 significant figures |
Temperature (Metric) Operating | -5°C to +95°C |
Unit Size | Each |
Description | CPVC |
Range | 0 to 5000 µS/cm; 0 to 18.2 MW·cm |
Electrode | Two electrode measurement with 0.01, 0.1, and 1.0 cell constants |
Max. Pressure | 150psig (at 75°C); 150psig (at 95°C) |
Type | Conductivity Resistivity Sensors, CPVC |
For Use With (Equipment) | AquaSensors™ DataStick™ Analytical Measurement System |
Flow Rate | 10 ft./sec. |
Resolution | 4.5 significant figures |
Temperature (Metric) Operating | -5°C to +95°C |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CCPEEK | Each | PEEK | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CCCPVC | Each | CPVC | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
AquaSensors Conductivity/Resistivity DataStick:
Conductivity Sensor Head:
Communications Adapter:
AV38 Local Display/Controller:
Recommended for: