Thermo Scientific™

Nalgene™ Rigid Containment Bottles

Catalog number: CE-N3233-42
Have Questions?
Thermo Scientific™

Nalgene™ Rigid Containment Bottles

Catalog number: CE-N3233-42
Have Questions?
The Thermo Scientific™ Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. This enables you to choose the appropriate product necessary to suit your application—through the step-down process and into your clean room.
 
Catalog Number
CE-N3233-42
Unit Size
Case of 20
Capacity (Metric)
2 L
Shape
Square
Price (USD)
Full specifications
Capacity (Metric)2 L
Capacity (English)67.63 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI, USP 661, FDA 21 CFR 177.1315
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)265 mm
Product TypeStorage and Transport Bottle
Width (English)19.75 in.
Height (English)10.43 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size38 mm
Diameter (Metric)116 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 20
Catalog Number
Unit Size
Each
Capacity (Metric)
20 mL
Shape
Round
Price (USD)
Full specifications
Capacity (Metric)20 mL
Capacity (English)0.67 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeRound
Height (Metric)69 mm
Product TypeStorage and Transport Bottle
Width (English)11 in.
Height (English)2.72 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size20 mm
Diameter (Metric)30 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
Catalog Number
Unit Size
Each
Capacity (Metric)
125 mL
Shape
Square
Price (USD)
Full specifications
Capacity (Metric)125 mL
Capacity (English)4.23 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Low Particulate<1/3 USP <788> (Ultralow particulate)
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)104.6 mm
Product TypeStorage and Transport Bottle
Width (English)12.5 in.
Height (English)4.13 in.
Closure LinerLined
Closure MaterialPolycarbonate
Closure Size38 mm
Diameter (Metric)52.1 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
Catalog Number
Unit Size
Case of 35
Capacity (Metric)
1 L
Shape
Square
Price (USD)
Full specifications
Capacity (Metric)1 L
Capacity (English)33.8 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI, USP 661, FDA 21 CFR 177.1315
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)201 mm
Product TypeStorage and Transport Bottle
Width (English)20.5 in.
Height (English)7.91 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)98 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 35
Catalog Number
Unit Size
Case of 6
Capacity (Metric)
5 L
Shape
Square
Price (USD)
Full specifications
Capacity (Metric)5 L
Capacity (English)169 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)299 mm
Product TypeStorage and Transport Bottle
Width (English)14.3 in.
Height (English)11.77 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)166 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 6
Catalog Number
Unit Size
Each
Capacity (Metric)
10 L
Shape
Square
Price (USD)
Full specifications
Capacity (Metric)10 L
Capacity (English)338.1 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)337 mm
Product TypeStorage and Transport Bottle
Width (English)16 in.
Height (English)13.27 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)255 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeCapacity (Metric)ShapePrice (USD)
CE-N3233-42Full specifications
Case of 202 LSquareRequest A Quote
Capacity (Metric)2 L
Capacity (English)67.63 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI, USP 661, FDA 21 CFR 177.1315
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)265 mm
Product TypeStorage and Transport Bottle
Width (English)19.75 in.
Height (English)10.43 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size38 mm
Diameter (Metric)116 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 20
CE-N3500-20Full specifications
Each20 mLRoundRequest A Quote
Capacity (Metric)20 mL
Capacity (English)0.67 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeRound
Height (Metric)69 mm
Product TypeStorage and Transport Bottle
Width (English)11 in.
Height (English)2.72 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size20 mm
Diameter (Metric)30 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
CE-N3030-42Full specifications
Each125 mLSquareRequest A Quote
Capacity (Metric)125 mL
Capacity (English)4.23 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Low Particulate<1/3 USP <788> (Ultralow particulate)
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)104.6 mm
Product TypeStorage and Transport Bottle
Width (English)12.5 in.
Height (English)4.13 in.
Closure LinerLined
Closure MaterialPolycarbonate
Closure Size38 mm
Diameter (Metric)52.1 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
CE-N3120-42Full specifications
Case of 351 LSquareRequest A Quote
Capacity (Metric)1 L
Capacity (English)33.8 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI, USP 661, FDA 21 CFR 177.1315
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)201 mm
Product TypeStorage and Transport Bottle
Width (English)20.5 in.
Height (English)7.91 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)98 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 35
CE-N3405-42Full specifications
Case of 65 LSquareRequest A Quote
Capacity (Metric)5 L
Capacity (English)169 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)299 mm
Product TypeStorage and Transport Bottle
Width (English)14.3 in.
Height (English)11.77 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)166 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeCase of 6
CE-N3410-42Full specifications
Each10 LSquareRequest A Quote
Capacity (Metric)10 L
Capacity (English)338.1 oz.
ColorClear
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
MaterialPolycarbonate
SterilitySterile
Temperature Range (Metric)-100°C to 100°C
Liner MaterialSilicone
Certifications/ComplianceUSP Class VI
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Height (Metric)337 mm
Product TypeStorage and Transport Bottle
Width (English)16 in.
Height (English)13.27 in.
Closure LinerLined
Closure MaterialPolypropylene
Closure Size48 mm
Diameter (Metric)255 mm
Closure TypeCap
Through the Closure DispensingNo
Unit SizeEach
Showing 6 of 6

Multiple tiers

  • Platinum certified clean containers are certified to contain less than 1/3 of the allowable particulate limits specified in USP <788>
  • Certified clean containers meet USP <788> low particulate requirements

Multiple resins available

  • Polycarbonate (PC)
  • Polyethylene terephthalate (PETG)
  • High-density polyethylene (HDPE)

Sterile, ready-to-use products

  • Robust triple-layer packaging for your step-down process
    • 2 heat-sealed bags create dual sterile barriers
    • 1 carton liner
    • Double-walled cartons
    • Improved labeling
  • Produced in clean room (ISO class 7)
  • Certified leakproof* to protect valuable contents

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text