Hastelloy PT1000 External Probe
Hastelloy PT1000 External Probe
Thermo Scientific™
Hastelloy PT1000 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Have Questions?
Catalog NumberType
CIC0001449Probe
Catalog number CIC0001449
Price (USD)
125.90
Online Exclusive
146.00
Save 20.10 (14%)
Each
In stock
Add to cart
Type:
Probe
Price (USD)
125.90
Online Exclusive
146.00
Save 20.10 (14%)
Each
Add to cart
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Specifications
DescriptionHastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
DisplayDigital
For Use With (Equipment)Cimarec and Supernuova Hotplates and Hotplate Stirrers
Temperature (Metric)0°C to 400°C
TypeProbe
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link