Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Calibration | 1.5 mg/L primary chlorine standard |
Description | AQUAfast chlorine primary standard kit to prepare 1.5 mg/L NIST-traceable chlorine standard |
For Use With (Equipment) | AQ3170 Chlorine Colorimeter |
Includes | Zero chlorine demand water, ampoule with primary chlorine standard and instructions |
Unit Size | Each |
Accuracy | ±0.05 mg/L (0.02 to 1.00 mg/L); ±0.10 mg/L (1.01 to 2.00 mg/L); ±0.20 mg/L (2.01 to 3.00 mg/L); ±0.30 mg/L (3.01 to 4.00 mg/L); ±0.40 mg/L (4.01 to 8.00 mg/L) |
Backlight Option | Yes |
Battery Life | 17 hours operating time or 5,000 measurements in continuous mode when display backlight is off |
Calibration | User and factory calibration functions |
Certifications/Compliance | CE |
Data Storage | 16 with date/time stamp |
Description | AQUAfast AQ3170 Chlorine Colorimeter |
Battery Type | 4 x AAA |
Display | Backlit LCD |
Dimensions (L x W x H) | 155 x 75 x 35 mm |
IP Rating | IP67 |
Light Source | LED |
Product Line | AQUAfast |
Type | Chlorine Colorimeter |
For Use With (Application) | Reagents: AC4P71, AC4P72, AQ250F, AQ250T; Vials: AC2V24, AC2V10; Standards: CLSK100, CLSK200 |
Includes | Colorimeter with free and total chlorine powder packs (100 each), sample vials with lids, field case, batteries and literature |
Packaging | Powder packs, bulk powder dispensers |
Range | Low Range: 0.02 to 2.00 mg/L; High Range: 0.10 to 8.00 mg/L |
Test Method | Dedicated Chlorine Colorimeter |
Warranty | 2 Years |
Wavelength | 530 nm |
Wavelength Accuracy | ± 1 nm |
Weight (Metric) | 260 g (With batteries) |
Unit Size | Each |
Description | AQUAfast chlorine secondary standards (0.0, 0.20 and 1.0 mg/L chlorine) for AQ3170 colorimeter |
For Use With (Equipment) | AQ3170 Chlorine Colorimeter |
Includes | Sealed vials with 0.0, 0.2 and 1.0 mg/L secondary chlorine standards and case |
Unit Size | Each |
Description | AQUAfast 10mm short path-length plastic vials for high level chlorine, 12 pack |
For Use With (Equipment) | AQ3170 Chlorine Colorimeter |
Includes | 12 pack of 10 mm plastic short path-length vials |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CLSK100 | Each | AQUAfast chlorine primary standard kit to prepare 1.5 mg/L NIST-traceable chlorine standard | 40.41 Online price 44.90 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ3170 | Each | AQUAfast AQ3170 Chlorine Colorimeter | 583.20 Online price 648.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CLSK200 | Each | AQUAfast chlorine secondary standards (0.0, 0.20 and 1.0 mg/L chlorine) for AQ3170 colorimeter | 172.00 Online price 200.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC2V10 | Each | AQUAfast 10mm short path-length plastic vials for high level chlorine, 12 pack | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|