Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detect and confirm the presence of members of coli-aerogenes group in water, wastewater, foods, milk, and dairy products with Thermo Scientific™ Oxoid™ Brilliant Green Bile Broth 2% (Dehydrated). The medium has been formulated to select for coliform bacteria which are recognised by rapid production of gas through lactose fermentation. Bile salts and brilliant green in the medium inhibit the Gram-positive and Gram-negative bacteria respectively.
Certifications/Compliance | Industrial Reference: AFNOR, AOAC, BSI, IDF, ISO, NMKL. |
Description | Brilliant Green Bile 2% Broth |
Form | Powder |
Product Type | Dehydrated Culture Media |
Quantity | 500 g |
Yield | for 12.5L medium |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CM0031B | Each | 500 g | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Brilliant Green Bile Broth 2% was first described by Durham and Schoenlein1 to selectively recover organisms of the coli-aerogenes group. The concentration of the components of the medium is optimized so that anaerobic lactose fermenters are inhibited without the coliforms being affected.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.