Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Sabouraud Dextrose Agar |
Form | Powder |
Product Type | Agar |
Quantity | 5 kg |
Shelf Life | 5 Years |
Unit Size | Each |
Description | Sabouraud Dextrose Agar |
Form | Powder |
Product Type | Agar |
Quantity | 500 g |
Shelf Life | 5 Years |
Unit Size | Each |
Description | Sabouraud Dextrose Agar |
Form | Powder |
Product Type | Agar |
Quantity | 2.5 kg |
Shelf Life | 5 Years |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Quantity | Content And Storage | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CM0041T | Each | Sabouraud Dextrose Agar | 5 kg | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CM0041B | Each | Sabouraud Dextrose Agar | 500 g | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CM0041R | Each | Sabouraud Dextrose Agar | 2.5 kg | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|