Liver Broth Culture Medium (Dehydrated)
Liver Broth Culture Medium (Dehydrated)
Liver Broth Culture Medium (Dehydrated)
Liver Broth Culture Medium (Dehydrated)
Thermo Scientific™
Liver Broth Culture Medium (Dehydrated)
Cultivate and enumerate saccharolytic or putrefactive mesophilic and thermophilic anaerobes from the food samples with this medium.
Have Questions?
Catalog NumberQuantity
CM0077B500 g
Catalog number CM0077B
Price (USD)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Cultivate and enumerate saccharolytic or putrefactive mesophilic and thermophilic anaerobes from the food samples with Thermo Scientific™ Oxoid™ Liver Broth Culture Medium (Dehydrated). The medium contains liver particles which support luxurious growth for anaerobes.
Liver Broth Culture Medium is a liver infusion medium, containing liver particles, for the examination of foods for saccharolytic or putrefactive mesophilic and thermophilic anaerobes. The medium is also recommended for the maintenance of aerobes and anaerobes in pure culture. Gillespie in communication with Scarr1 recommended Liver Broth for the examination of canners’ sugar for hydrogen swells caused by thermophilic anaerobes (Clostridium thermosaccharolyticum).
A 20% w/v solution of the sugar is steamed for 30 minutes to destroy vegetative forms and inoculated into Liver Broth sealed with agar. The standard proposed was a maximum of one positive tube in six, with 20mL inocula incubated for 72 hours at 56°C.
This medium should be made up only when required for use. Storage of the reconstituted medium is not recommended because air may be absorbed and the re-steaming necessary for the restoration of anaerobic conditions darkens the medium. Liver broth is not an homogeneous medium, consisting of a layer of liver particles and a cloudy supernatant. Growth produces an obvious increase in turbidity and some organisms (e.g. Clostridium thermosaccharolyticum ) also produce gas which often pushes the agar plug towards the top of the tube. Some organisms also digest the solid liver tissue.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
1. Scarr M. Pamela (1958) DSIR, Proc. 2nd Internat. Symp. Food Microbiol. 1957, HMSO London, pp. 29-33.
Specifications
DescriptionLiver Broth Culture Medium (Dehydrated)
Quantity500 g
Shelf Life5 Years
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
10°C to 30°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection