Search Thermo Fisher Scientific
Isolate and identify Salmonella and Shigella species from clinical or food samples with this selective medium.
Catalog Number | Quantity |
---|---|
CM0099B | 500 g |
CM0099K | 25 kg |
Isolate and identify Salmonella and Shigella species from clinical or food samples with Thermo Scientific™ Oxoid™ SS Agar (Dehydrated). Differentiation of Salmonella and Shigella is achieved by incorporation of lactose in the medium. Organisms which ferment lactose produce acid which, in the presence of the neutral red indicator, results in the formation of red colonies. Salmonella, Shigella, and other non-lactose-fermenting organisms appear as transparent or straw coloured colonies on SS Agar.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
SS Agar is a differential, selective medium for the isolation of Shigella and Salmonella species from pathological specimens, suspected foodstuffs etc.
Thiosuplate in combination with iron acts as an indicator of sulphide production, which is indicated by blackening in the centers of the colonies.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Share catalog number, name or link