Thermo Scientific™
Antibiotic Medium No. 3 Assay Broth (Dehydrated)
Catalog number: CM0287B
Have Questions?
Thermo Scientific™
Antibiotic Medium No. 3 Assay Broth (Dehydrated)
Catalog number: CM0287B
Have Questions?
Carry out serial dilution assay of penicillin and other antibiotics with Thermo Scientific™ Oxoid™ Antibiotic Medium No. 3 Assay Broth (Dehydrated). Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Catalog Number
CM0287B
Unit Size
Each
Quantity
500 g
Price (USD)
Full specifications
DescriptionAntibiotic Medium No. 3
FormPowder
Product TypeDehydrated Culture Media
Quantity500 g
YieldFor 28.6L medium
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
CM0287BFull specifications
Each500 gRequest A Quote
DescriptionAntibiotic Medium No. 3
FormPowder
Product TypeDehydrated Culture Media
Quantity500 g
YieldFor 28.6L medium
Unit SizeEach
Showing 1 of 1
Antibiotic Medium No. 3 Assay Broth is used as the broth medium in turbidimetric or serial dilution assay of a wide variety of antibiotics with Staphylococcus aureus.
Taking advantage of modern technology, this medium is based on the original `Penassay Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand