Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
產品號碼 | Quantity |
---|---|
CM0331B | 500 g |
CM0331T | 5 kg |
CM0331K | 25 kg |
CM0331R | 2.5 kg |
Cultivate and isolate fastidious microorganisms with clearly visible hemolytic reactions with Thermo Scientific™ Oxoid™ Columbia Blood Agar Base (Dehydrated) when used with whole defibrinated blood (SR0050). Columbia Blood Agar Base is an ideal base medium for preparation of blood agar, chocolate agar and for preparation of various selective and identification media.
Grow fastidious organisms with rapid production of large colonies, clearly defined zones of hemolysis and good colonial differentiation for all-around performance with Columbia Blood Agar when used with whole defibrinated blood.
Traditionally, blood agar has been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and the second in clearly defined zones of hemolysis and good colonial differentiation. This formulation combines the virtues of both for optimal performance1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Ellner P.D., Stoessel C.J., Drakeford E. and Vasi F. (1966) Tech. Bull. Reg. Med. Techn. 36. No. 3, reprinted in Amer. J. Clin. Path. (1966) 45. 502-504.
Nonselective, general purpose medium which may be enriched with blood or supplements for the isolation and culture of microrganisms from clinical and non-clinical samples.
分享目錄編號、產品名稱或連結。