MRS Broth (Dehydrated)
MRS Broth (Dehydrated)
Thermo Scientific™
MRS Broth (Dehydrated)
Isolate and enumerate lactic acid bacteria from food with this non-selective medium.
Have Questions?
Catalog NumberQuantity
CM0359B500 g
Catalog number CM0359B
Price (USD)
-
Request A Quote
Quantity:
500 g
Isolate and enumerate lactic acid bacteria from food with Thermo Scientific™ Oxoid™ MRS (DeMan, Rogosa and Sharpe) Broth (Dehydrated). This non-selective medium is designed to support profuse growth of lactic acid bacteria.
Use MRS Broth for enumeration and isolation of mesophilic lactic acid bacteria from food samples. MRS Broth may also be used for tests in the identification of lactobacilli, such as temperature dependence, growth in 4% NaCl, growth in 0.4% Teepol, etc. as recommended by Sharpe, Fryer, and Smith1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References:
  1. Sharpe M. Elisabeth, Fryer T. F. and Smith D. G. (1966) Identification of the Lactic Acid Bacteria in Identification Method for Microbiologists Part A (Gibbs B. M. and Skinner F. A. eds.) London and New York, Academic Press. Pages 65-79.
Specifications
Certifications/ComplianceIndustrial Reference: AFNOR, IDF, NMKL.
DescriptionMRS Broth
Quantity500 g
YieldFor 9.6L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Share catalog number, name or link