Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
CM0391B | 500 g |
CM0391T | 5 kg |
Oxoid Thioglycollate Broth, Alternative is a non-viscous thioglycollate medium is used for the culture of turbid biological products.
Share catalog number, name or link