Search Thermo Fisher Scientific
Catalog Number | Form | Quantity |
---|---|---|
CM0395B | Powder | 500 g |
Selenite Broth Base contains lactose as a source of fermentable carbohydrate which helps to prevent any rise in pH value during incubation, thereby preventing any loss of selective activity of selenite.
Use Selenite Broth Base for enrichment and isolation of Salmonella from feces, urine, water, foods, and materials of sanitary importance.
Klett1 first demonstrated the selective inhibitory effects of selenite, and Guth2 used it to isolate Salmonella typhi. It was 20 years later before Leifson3 fully investigated selenite and promoted wide use of the medium.
Proteus and Pseudomonas species appear to be resistant to its effects4. Lactose is added as a fermentable carbohydrate to prevent a rise in pH value during incubation because any increase in pH will reduce the selective activity of selenite. The fact that Proteus and Pseudomonas species do not ferment lactose may explain why they escape inhibition.
To minimise any possible risk of teratogenicity to laboratory workers, the sodium biselenite must be added as a solution to this medium. Robertson5 reported miscarriages and possible teratogenic effects on pregnant laboratory assistants which may have been caused by ingested sodium biselenite. This substance has, therefore, been removed from the Oxoid powdered medium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References:
An enriched medium for the isolation of Salmonella species from faecal and urine samples. Can also be used for testing water and food samples.
Share catalog number, name or link