Oxoid™ SS Agar Modified
Oxoid™ SS Agar Modified
Thermo Scientific™
Oxoid™ SS Agar Modified
Selectively isolate, differentiate Shigella and Salmonella species from pathological specimens and suspected foodstuffs with this medium.
Have Questions?
Catalog NumberQuantity
CM0533B500 g
Catalog number CM0533B
Price (USD)
-
Request A Quote
Quantity:
500 g
Selectively isolate, differentiate Shigella and Salmonella species from pathological specimens and suspected foodstuffs with Thermo Scientific™ Oxoid™ SS Agar Modified (Dehydrated). SS Agar Modified is designed to overcome the difficulties faced by SS agar. The improved formulation includes an alteration to the bile salt mixture, peptone, and pH value, improving its performance in the growth of shigellae and improved colony size for salmonellae with improved blackening at the center.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. SS Agar Modified is ideal for screening samples containing mixed flora and suspected of harboring enteric pathogens, e.g., medical specimens or food products.
  • Easy-to-read: shigellae form transparent colonies while salmonellae form transparent colonies with black centers
  • Selective: selective inhibitory components brilliant green, bile salts, thiosulphate, and citrate inhibit Gram-positive organisms and non-target coliforms.
Although widely used, SS Agar has been criticized because of excessive inhibition of Shigella species.
Investigation has shown that modification to the formulation by alterations to the bile salt mixture, peptone and pH value considerably improve its performance in the growth of shigellae without too much increased growth of commensal organisms.
Salmonella colonies are also larger with improved blackening at the center. The change in formulation has reduced the number of g/L from 63g to 57g.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSS Agar Modified
Quantity500 g
YieldFor 8.8L medium
FormPowder
Product TypeAgar
Unit SizeEach
Intended use statement 
A differential selective medium for the isolation of Salmonella and Shigella species from faecal samples. Can also be used for testing food samples
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link