Perfringens TSC Agar Base (Dehydrated)
Perfringens TSC Agar Base (Dehydrated)
Thermo Scientific™
Perfringens TSC Agar Base (Dehydrated)
Selectively isolate and presumptively identify Clostridium perfringensin food samples with this basal medium.
Have Questions?
Catalog number CM0587B
Price (USD)/ Each
-
Request A Quote
Quantity:
500 g
Selectively isolate and presumptively identify Clostridium perfringens in food samples with Thermo Scientific™ Oxoid™ Perfringens TSC Agar Base (Dehydrated). The medium can either be used on its own or with selective agents to make Tryptose Sulphite (TS) Agar, Tryptose Sulphite Cycloserine (TSC)1 agar or Shahadi Ferguson Perfringens (SPF)2 agar for the presumptive identification and enumeration of Clostridium perfringens.
Meat peptone, soy peptone and yeast extracts provide essential nutrients and vitamins for the development of clostridia. Sodium metabisulphite and ferric ammonium citrate act as an indicator of sulphide reduction, indicated by black coloured colonies.
Some strains of Clostridium perfringens may produce an opaque zone around the colony due to lecithinase activity, but this is not considered to be universal for all Clostridium perfringens strains after overnight incubation3, and both black, lecithinase positive and black, lecithinase negative colonies should be considered as presumptive Clostridium perfringens on TSC Agar and confirmatory tests carried out.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
1. Harmon S. M., Kauttar D. A. and Peeler J. T. (1971) Appl. Micobiol. 22. 688-692.
2. Shahidi S. A. and Ferguson A. R. (1971) Appl. Microbiol. 21. 500-506
3. Hauschild A. H. W. and Hilsheimar R. (1974) Appl. Microbiol. 27. 78-82
Specifications
DescriptionPerfringens Agar Base (TSC/SFP)
FormPowder
Product TypeAgar
Quantity500 g
YieldFor 10.9L medium
Unit SizeEach