Rogosa Agar (Dehydrated)
Rogosa Agar (Dehydrated)
Rogosa Agar (Dehydrated)
Rogosa Agar (Dehydrated)
Thermo Scientific™
Rogosa Agar (Dehydrated)
Selectively isolate and enumerate lactobacilli from dairy and other pathological samples with this medium.
Have Questions?
Catalog NumberQuantity
CM0627B500 g
Catalog number CM0627B
Price (EUR)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Selectively isolate and enumerate Lactobacilli with Thermo Scientific™ Oxoid™ Rogosa Agar (Dehydrated). The medium has given excellent results when used in quantitative and qualitative studies of lactobacilli in feces, saliva and mouth rinses, and in dairy products.
Rogosa Agar, a modification of the medium described by Rogosa et al.1, is a selective medium for the isolation and enumeration of lactobacilli. High acetate concentration and low pH of the medium effectively suppress other bacteria, but also many strains of other lactic acid bacteria. The modification of the pH to 6.2 instead of 5.5 alters the selectivity of the medium for the whole group of lactic acid bacteria2,3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use
For Laboratory Use
Specifications
DescriptionRogosa Agar
Quantity500 g
YieldFor 6.1L medium
FormPowder
Product TypeAgar
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6233012Certificate of AnalysisMar 07, 2025CM0627B
6137391Certificate of AnalysisFeb 01, 2025CM0627B
3725350Certificate of AnalysisOct 11, 2023CM0627B
3645945Certificate of AnalysisApr 14, 2023CM0627B
3540111Certificate of AnalysisSep 13, 2022CM0627B
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection