Rappaport-Vassiliadis Enrichment Broth (Dehydrated)
Rappaport-Vassiliadis Enrichment Broth (Dehydrated)
Thermo Scientific™
Rappaport-Vassiliadis Enrichment Broth (Dehydrated)
Selectively enrich Salmonella spp. from food and environmental specimens with this selective medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0669B500 g
CM0669R2.5 kg
Catalog number CM0669B
Price (USD)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Selectively enrich Salmonella spp. from food and environmental specimens with Thermo Scientific™ Oxoid™ Rappaport-Vassiliadis Enrichment Broth (Dehydrated). Rappaport-Vassiliadis Enrichment Broth selects for Salmonella species by exploiting their characteristics when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements1.
Salmonella are a group of closely related rod-shaped, Gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food. The most common symptoms of Salmonella infection are diarrhea, fever, abdominal cramps, and vomiting.
Use Rappaport-Vassiliadis Enrichment Broth for selective enrichment of Salmonella species food and environmental samples.
  • Selective: malachite green, low pH, and high magnesium chloride concentration inhibit many Gram-positive bacteria, while selectively enriching for salmonellae.
  • Superior performance: superior performance to Selenite Enrichment Broth and Tetrathionate Broth for the enrichment of salmonellae with the exception of Salmonella Typhi1.
Rappaport-Vassiliadis (RV) Enrichment Broth is similar to that described by Vassiliadis et al.3 except that the peptone used is soya peptone, which has been reported to enhance the growth of salmonellae1,2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
DescriptionRappaport-Vassiliadis Enrichment Broth
Quantity500 g
YieldFor 16.7L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6271369Certificate of AnalysisJun 19, 2025CM0669B
6271369Certificate of AnalysisJun 18, 2025CM0669R
6210046Certificate of AnalysisFeb 01, 2025CM0669R
6157667Certificate of AnalysisDec 09, 2024CM0669B
6174059Certificate of AnalysisNov 01, 2024CM0669B
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection