Listeria Selective Oxford Agar Base (Dehydrated)
Listeria Selective Oxford Agar Base (Dehydrated)
Thermo Scientific™
Listeria Selective Oxford Agar Base (Dehydrated)
Isolate and detect Listeria monocytogenes from pathological, food and environmental samples with this diagnostic medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0856B500 g
CM0856R2.5 kg
Catalog number CM0856B
Price (USD)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Isolate and detect Listeria monocytogenes from, food and environmental samples with the diagnostic Thermo Scientific™ Oxoid™ Listeria Selective Oxford Agar Base (Dehydrated) when used with Modified Listeria Selective Supplement (Oxford) SR0206. The medium is based on the formulations described by Curtis et al.1 This medium is the specified plating medium in the FDA/BAM isolation procedure2 and in standardized testing methods of other national and international bodies3.
Foodborne infection by Listeria monocytogenes has prompted increased concern for detecting this organism in foods, in the environment and in pathological specimens from both human and animal subjects. The ability to isolate the organism has been impeded in the past by lack of an effective selective medium, as Listeria monocytogenes can be easily and completely overgrown by competing flora.
Use Listeria Selective Oxford Agar Base for selective isolation and detection of Listeria monocytogenes from food and environmental samples.
  • Easy-to-read: Listeria monocytogenes form brown colored colonies with black zones due to aesculin hydrolysis.
  • An FDA/BAM specified plating medium for Listeria monocytogenes isolation3.
The Listeria Selective Oxford Agar Base utilizes the selective inhibitory components and the indicator system―aesculin and ferrous iron for the isolation or differentiation of Listeria monocytogenes. Listeria monocytogenes hydrolyzes the aesculin, producing black zones around the colonies due to the formation of black iron phenolic compounds derived from the aglucon. Gram-negative bacteria are completely inhibited. Most unwanted Gram-positive species are suppressed, but some strains of enterococci grow poorly and exhibit a weak aesculin reaction, usually after 40 hours incubation. Some staphylococci may grow as aesculin-negative colonies.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Certifications/ComplianceIndustrial Reference: AFNOR, IDF, ISO, USDA.
DescriptionListeria Selective Agar Base (Oxford)
Quantity500 g
YieldFor 8.0L medium
FormPowder
Product TypeAgar
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6251576Certificate of AnalysisJun 14, 2025CM0856B
6220656Certificate of AnalysisFeb 04, 2025CM0856B
3806823Certificate of AnalysisJul 10, 2024CM0856B
6128270Certificate of AnalysisJun 28, 2024CM0856B
3705193Certificate of AnalysisAug 18, 2023CM0856B
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection