Rappaport-Vassiliadis Soya Peptone Broth (Dehydrated)
Rappaport-Vassiliadis Soya Peptone Broth (Dehydrated)
Thermo Scientific™

Rappaport-Vassiliadis Soya Peptone Broth (Dehydrated)

Selectively enrich Salmonella spp. from food and environmental specimens with this selective medium.
Have Questions?
更改视图buttonViewtableView
货号数量
CM0866B500 g
CM0866R2.5 kg
货号 CM0866B
价格(CNY)
-
申请报价
数量:
500 g
物流信息
常规产品: 发货后, 北上广深及省会城市通常为 1-2天,二线城市通常 2-3天,三线以及偏远地区通常 4天,周末以及节假日顺延。
空运受限产品: 发货后, 北上广深及省会城市通常为 2-4天,二线城市 通常3-5天,三线以及偏远地区通常 6-10天,周末以及节假日顺延。
现货中心: 此类产品将由您身边的现货中心极速配送。

Selectively enrich Salmonella spp. from food and environmental specimens with Thermo Scientific™ Oxoid™ Rappaport-Vassiliadis Soya Peptone Broth (Dehydrated). Rappaport- Vassiliadis Soya Peptone Broth selects for Salmonella species by exploiting their characteristics when compared with other Enterobacteriaceae, such as the ability to survive at relatively high osmotic pressure, multiply at relatively low pH values, be relatively more resistant to malachite green, and have relatively less demanding nutritional requirements1.

Use Rappaport-Vassiliadis Soya Peptone Broth for selective enrichment of Salmonella spp. from food and environmental specimens.

  • Selective: malachite green, low pH, and high magnesium chloride concentration inhibit many Gram-positive bacteria, while selectively enriching for salmonellae.
  • Superior performance to Selenite Enrichment Broth and Tetrathionate Broth for the enrichment of salmonellae with the exception of Salmonella typhi1

RVS Broth is based on the revised formulation described by van Schothorst et al.3, and is recommended as the selective enrichment medium for the isolation of salmonellae from food and environmental specimens. It can also be used to isolate salmonellae from human feces without the need for pre-enrichment. It is a modification of the Rappaport Vassiliadis (RV) Enrichment Broth described earlier by van Schothorst and Renauld4. The modifications to their earlier formula are:

  • The addition of dipotassium hydrogen phosphate to buffer the medium so that the pH is maintained during storage of the prepared broth.
  • Clarifying the optimum concentration of magnesium chloride 6H2O.
  • The two modifications are said to enhance the reliability of the enrichment broth1. Peterz et al.3 have also highlighted the importance of the concentration of magnesium chloride in the final medium.

    RVS Peptone Broth is based on the revised formulation and is recommended as the selective enrichment medium for the isolation of salmonellae from food and environmental specimens. It can also be used to isolate salmonellae from human feces without the need for pre-enrichment.

    Not all products are available for sale in all territories. Please inquire.

    Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

    规格
    认证/合规行业参考:ISO、NMKL。
    描述Rappaport Vassiliadis 大豆 (RVS) 蛋白胨肉汤(干粉)
    数量500 g
    产量适用于 18.7 L 培养基
    形式粉末
    产品类型干粉培养基
    Unit SizeEach

    分享货号、产品名称或链接