Azide Dextrose Broth (Dehydrated)
Azide Dextrose Broth (Dehydrated)
Thermo Scientific™
Azide Dextrose Broth (Dehydrated)
Detect enterococci in water and sewage samples with this selective medium.
Have Questions?
Catalog NumberQuantity
CM0868B500 g
Catalog number CM0868B
Price (CLP)
-
Quantity:
500 g
Ask our AI about this Product
Detect enterococci in water and sewage samples with Thermo Scientific™ Oxoid™ Azide Dextrose Broth (Dehydrated). A blend of peptone and glucose in the medium renders Azide Dextrose Broth highly nutritious, and sodium chloride maintains the osmotic equilibrium. Sodium azide acts as an inhibitor of Gram-negative organisms1,2,3 and the concentration of sodium azide used in the medium provides optimum protection for the enterococci while largely suppressing the Gram-negative flora. The phosphate buffer system controls pH.
Enterococci are part of the normal intestinal flora of human and animals. They have been long recognized as important human pathogens and are becoming increasingly so. The presence of enterococci serves as an indicator of fecal contamination. Enterococci are better indicators than Escherichia coli of sewage pollution in chlorinated waters because they have a greater resistance to chlorine.
Azide Dextrose broth is recommended for the detection of enterococci in water and sewage4.
  • Selective: sodium azinde in the medium acts as an inhibitor of Gram-negative organisms while providing optimum protection to enterococci.
  • Recommended for the quantitative determination of enterococci in water, sewage, foods and other materials suspected of contamination with sewage1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
General References:
  1. Mallmann W. L. and Seligmann E. B. (1950) Am. J. Public Health 40. 286.
  2. Edwards S. J. (1933) J. Comp. Path. Therap. 46. 211.
  3. Hartman G. (1937) Milchw. Forsch. 18. 166.
  4. Greenberg A. E. et al (ed). (1998) Standard Methods for the Examination of Water and Wastewater, 20th ed. APHA, Washington, D.C.
Specifications
DescriptionAzide Dextrose Broth
Quantity500 g
YieldFor 14.0L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6262743Certificate of AnalysisJun 14, 2025CM0868B
6235490Certificate of AnalysisMar 21, 2025CM0868B
6180182Certificate of AnalysisNov 13, 2024CM0868B
6139803Certificate of AnalysisSep 12, 2024CM0868B
6152335Certificate of AnalysisSep 12, 2024CM0868B
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection