Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Oxoid Baird-Parker RPF Agar Base is a selective medium for the isolation and enumeration of coagulase positive staphylococci from food.
Certifications/Compliance | Industrial Reference: ISO, NMKL. |
Description | Baird-Parker Agar Base, RPF |
Form | Powder |
Product Type | Agar |
Quantity | 500 g |
Yield | For 7.1L medium |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CM0961B | Each | 500 g | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|