Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detect Escherichia coli O157 with selective and differential Thermo Scientific™ Oxoid™ Sorbitol MacConkey Agar (SMAC) incorporating the chromogen 5-bromo-4-chloro-3-indolyl-b -D-glucuronide (BCIG) (Dehydrated).
SMAC with BCIG combines two different screening mechanisms for the detection of Escherichia coli O157, the failure to ferment sorbitol and the absence of b-glucuronidase activity. In a study with artificially contaminated meat samples, SMAC with BCIG reduced the number of false suspect Escherichia coli O157 colonies by 36%.1
Description | Sorbitol MacConkey Agar w/BCIG |
Form | Powder |
Product Type | Agar |
Quantity | 500 g |
Yield | for 9.7L medium |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CM0981B | Each | 500 g | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Enterohemorrhagic strains of Escherichia coli and in particular the serotype O157:H7 have presented an increasing concern among food and clinical microbiologists since their role as a causative agent in two major food related outbreaks of hemorrhagic colitis was established in 1982. Although a variety of serotypes are known to produce verotoxins; a three-year study in England and Wales showed that 99% of isolates from patients suffering diarrhoea, bloody diarrhoea or hemolytic uraemic syndrome (HUS) belonged to serogroup O1571.
The intestinal tract of ruminants is the prime reservoir of Escherichia coli O157 and other enterohemorrhagic Escherichia coli (EHEC) strains, therefore meats derived from cattle, sheep, goat and deer can be expected to be contaminated. Foods implicated in human illness related to Escherichia coli O157 include meats, dairy products, vegetables, salads, apple juice and water2.
The Sorbitol MacConkey Agar (SMAC) with BCIG formulation includes 5-bromo-4-chloro-3-indolyl- b-D-glucuronide (BCIG) as a substrate for b-glucuronidase, an enzyme which is not usually present in Escherichia coli O157 strains. The non-sorbitol-fermenting and b-glucuronidase-negative Escherichia coli O157 will appear as straw-coloured colonies. Organisms with b-glucuronidase activity will cleave the substrate and release indoxyl (or halogenated indoxyl) which is rapidly oxidized to the insoluble indigo or its analogue, leading to a distinct blue-green coloration of the colonies3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
1. Thomas, A., Cheasty, T., Frost, J. A., Chart, H., Smith, H. R. and Rowe, B. (1996) Epidemiol Infect. 117, 1-10.
2. Okrend, A. J. G., Rose, B. E. and Lattuada, C. P. (1990) J. Food Prot. 53, 941-943.
3. Desmarchelier, P. M. and Grau, F. H. (1997) Escherichia coli. In: Foodborne Microorganisms of Public Health Significance. 5th Edition. pp.231-264. A. D. Hocking (Ed.). AIFST (NSW Branch) Food Microbiology Group, Australia