Membrane Lactose Glucoronide Agar (MLGA)
Membrane Lactose Glucoronide Agar (MLGA)
Membrane Lactose Glucoronide Agar (MLGA)
Membrane Lactose Glucoronide Agar (MLGA)
Thermo Scientific™
Membrane Lactose Glucoronide Agar (MLGA)
Detect, enumerate, and differentiate Escherichia coli and other coliforms by single membrane filtration with this selective and differential medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM1031T5 kg
CM1031B500 g
Catalog number CM1031T
Price (USD)
-
Request A Quote
Quantity:
5 kg
Ask our AI about this Product
Detect, enumerate and differentiate Escherichia coli and other coliforms by single membrane filtration with Thermo Scientific™ Oxoid™ Membrane Lactose Glucuronide Agar (MLGA) (Dehydrated). The presence of lauryl sulphate and lactose with phenol red allow the selective isolation and differentiation of Escherichia coli and coliforms respectively.
Tests for Escherichia coli and coliforms provide a sensitive means for detection of fecal contamination of drinking water. They are regarded as the most important routine microbial examinations for assessing the effectiveness of water treatment and disinfection and for monitoring water quality in distribution1. Membrane Lactose Glucuronide Agar (MLGA) supports water testing utilising single membrane filtration.
  • Isolate coliforms effectively with MLGA using a single membrane filtration method.
  • Lauryl sulphate selects for coliforms by inhibiting Gram-positive organisms.
  • Reduced confirmatory tests for E. coli due to the specificity of the reactions in the medium.
  • Easy interpretation of the test results due to addition of chromogenic BCIG (5-bromo-4-chloro-3-indolyl-ß-D-glucuronide), E. coli appear as green colonies while other coliform bacteria form yellow colonies.
Identification of Escherichia coli and coliforms is facilitated through two biochemical reactions within the medium: Lactose fermentation is detected by the dye phenol red which gives yellow colonies when acid is produced. The chromogenic substrate BCIG is cleaved by the enzyme glucuronidase and produces a blue chromophore which builds up in the bacterial cells. Coliforms are lactose-positive so colonies will be yellow; Escherichia coli is both lactose-positive and possesses glucuronidase so will appear as green colonies.1 Strains of E. coli that fail to produce typical colonies on MLGA might produce lower concentrations of the enzyme β-D-glucuronidase.2
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
For Laboratory Use
For Laboratory Use
General References:
  1. The Environment Agency - Methods for Examination of Waters and Associated Material - The Microbiology of Drinking Water 2002.
  2. Letters in Applied Microbiology. Understanding the cause of false negative β-D-glucuronidase reactions in culture media containing fermentable carbohydrate. C.R. Fricker,P.S. Warden,B.J. Eldred
Specifications
DescriptionMembrane Lactose Glucuronide Agar (MLGA)(Dehydrated)
Quantity5 kg
Shelf Life3 Years
FormPowder
Product TypeAgar
Unit SizeEach
Contents & Storage
10°C to 30°C
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6248436Certificate of AnalysisJun 14, 2025CM1031T
6248436Certificate of AnalysisJun 14, 2025CM1031B
6235881Certificate of AnalysisMar 21, 2025CM1031T
6196876Certificate of AnalysisDec 07, 2024CM1031B
6196876Certificate of AnalysisDec 06, 2024CM1031T
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection