Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Carry out microbiological Media Fill Trials (MFT) in the pharmaceutical industry with gamma-irradiated Thermo Scientific™ Oxoid™ Cold Filterable Tryptone Soya Broth (Dehydrated). The highly nutritious general purpose medium conforms to the formulation stated for MFT in the European1, British2, US3 and Japanese4 Pharmacopoeias.
Cold Filterable Tryptone Soya Broth can be substituted for the powdered components that go into making aqueous drug products to provide an index of the level of microbial contamination occurring in aseptic manufacture.
Description | Cold Filterable Tryptone Soya Broth |
Form | Powder |
Product Type | Dehydrated Culture Media |
Quantity | 5 kg |
Yield | For 167.0L medium |
Unit Size | Each |
Description | Cold Filterable Tryptone Soya Broth |
Form | Powder |
Product Type | Dehydrated Culture Media |
Quantity | 500 g |
Yield | For 16.7L medium |
Unit Size | Each |
Description | Cold Filterable Tryptone Soya Broth |
Form | Powder |
Product Type | Dehydrated Culture Media |
Quantity | 25 kg |
Yield | For 833.0L medium |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
CM1065T | Each | 5 kg | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CM1065B | Each | 500 g | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
CM1065K | Each | 25 kg | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
‘Sterile for use’ liquid drugs often contain heat-sensitive components which means that terminal sterilisation by autoclaving is not an option. Sterilisation by filtering (for soluble liquids) followed by filling under aseptic conditions is the method for preparation of these types of drug. The purpose of Media Fill Trials is to provide a measure of the likelihood of microbiological contamination arising in a particular aseptic process.
Packs of Cold Filterable Tryptone Soya Broth have been given a sterilizing dose of gamma-irradiation (minimum 25KGy) validated as a lethal dose for all yeasts, molds, bacterial spores and mycoplasma.
The purpose of MFT is to provide a measure of the likelihood of microbiological contamination occurring in a particular aseptic process. Cold Filterable TSB can be used as a substitute for filter-sterilized drug products and is processed in a manner identical to that in which the product would be processed i.e. filtering, filling and closing. The medium is then incubated and the number of contaminated units is scored versus those that are un-contaminated. A decision following pre-determined guidelines can be made based on the proportion of contaminated units and the identity of the microorganisms recovered5.
Cold Filterable Tryptone Soya Broth may also be used to test aseptic production processes for solid presentations by adding the medium as a sterile liquid downstream of processing a placebo5.
For laboratory use, standard Thermo Scientific™ Oxoid™ Tryptone Soya Broth (CM0129) may be more appropriate.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
1. European Pharmacopoeia 4th Edition 2002
2. British Pharmacopoeia 2003
3. US Pharmacopoeia 27 NF22 2004
4. Japanese Pharmacopoeia XIV 2001
5. Microbiological Media Fills Explained by Nigel Halls, 2002. Sue Horwood Publishing Ltd., UK.