Brain Heart Infusion Broth (Dehydrated)
Brain Heart Infusion Broth (Dehydrated)
Thermo Scientific™
Brain Heart Infusion Broth (Dehydrated)
Cultivate streptococci, Neisseria and other fastidious organisms with this highly nutritious medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM1135B500 g
CM1135K25 kg
CM1135T5 kg
Catalog number CM1135B
Price (USD)
-
Request A Quote
Quantity:
500 g
Ask our AI about this Product
Cultivate streptococci, Neisseria and other fastidious organisms with Thermo Scientific™ Oxoid™ Brain Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms. It is recommended for blood culture work for the isolation and cultivation of pathogenic fungi.
Cultivate a wide variety of organisms with Brain Infusion Broth:
  • Use as a growth and suspension medium for staphylococci when testing coagulase production.
  • Add 10% defibrillated blood for isolation and cultivation of pathogenic fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionBrain Heart Infusion Broth
Quantity500 g
YieldFor 13.5L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
6246646Certificate of AnalysisJun 14, 2025CM1135B
6246645Certificate of AnalysisJun 14, 2025CM1135B
6224624Certificate of AnalysisApr 04, 2025CM1135B
6236076Certificate of AnalysisMar 25, 2025CM1135B
6235486Certificate of AnalysisMar 10, 2025CM1135B
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection