Hepatocyte Maintenance Supplement Packs allow you to supplement up to 500 ml of Williams Medium E without phenol red (orRead more
Have Questions?
Catalog Number
Quantity
CM4000
1 kit
Catalog number CM4000
Price (USD)
117.65
Online Exclusive
129.00
Save 11.35 (9%)
Each
In stock
Add to cart
Quantity:
1 kit
Price (USD)
117.65
Online Exclusive
129.00
Save 11.35 (9%)
Each
Add to cart
Hepatocyte Maintenance Supplement Packs allow you to supplement up to 500 ml of Williams Medium E without phenol red (or suitable alternative basal medium) for the purpose of incubating hepatocytes in suspension or plated cultures.
Hepatocyte Maintenance Supplement Packs contain: • Dexamethasone • A cocktail solution of penicillin-streptomycin, ITS+ (insulin, transferrin, selenium complex, BSA, and linoleic acid), GlutaMAX™, and HEPES
Note: Kit components may ship separately due to differences in storage temperatures.