Informational:Upgrade now to TrueCut Cas9 Protein v2 for the highest on-target editing efficiency.
Informational:Informational:100% knock-in using CRISPR-Cas9—try our new TrueTag Donor DNA Kits for fluorescent or epitope gene tagging.
Invitrogen™

Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Catalog number: CMAX00001
Have Questions?
Invitrogen™

Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Catalog number: CMAX00001
Have Questions?
Catalog Number
CMAX00001
Unit Size
0.1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.75 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.3 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
CMAX000010.1 mL
-
CMAX000080.75 mL
-
CMAX000030.3 mL
-
CMAX000151.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid nanoparticle transfection reagent for CRISPR-Cas9 protein delivery. It is a high-throughput-friendly, cost-effective alternative to electroporation.

Working together with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it provides:
• Demonstrated cleavage efficiency—tested in over 20 cell types including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and several others
• Low cell toxicity—less cells needed to initiate your experiment
• High throughput friendly—an ideal delivery solution for 96-well format
• Cost savings—whether cost-per-reaction or initial investment

Achieve high transfection efficiency in novel genome editing applications
Lipofectamine CRISPRMAX transfection reagent increases the likelihood of successful cleavage and recombination, especially when combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA. This ultimately maximizes the efficiency of performing genetic modifications.

Improve experimental outcomes using TrueCut Cas9 Protein v2
Unlike CRISPR plasmid or Cas9 mRNA, using Cas9 protein provides superior cleavage efficiency in primary cells and stem cells. It eliminates the need for transcription or translation of the payload, removes the risk of genomic integration, and is cell cycle independent.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Classification
Animal Origin-Free
Cell Type
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type
Protein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Product Line
CRISPRMAX™, Lipofectamine™
Product Type
Transfection Reagent
Quantity
0.1 mL
Serum Compatible
Yes
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice

Contents & Storage

Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text