Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | Range |
---|---|---|
CODH150 | AQUAfast Digestion Tubes, Range: 0 to 15000 mg/L O2, COD High Range | 0 to 15000 mg/L O2 |
AC2009 | AQUAfast fluoride SPADNS liquid reagent, 50 tests | 0.05 to 2mg/L F |
AC3001 | AQUAfast pH liquid dropper reagent, 30 tests | 6.5 to 8.4 pH |
AC4P54 | AQUAfast manganese LR reagent powder & liquid set, 50 tests | 0.01 to 0.7mg/L Mn |
ACD004 | AQUAfast total nitrogen LR digestion tubes, 50 pack | 0.5 to 25mg/L N |
ACD007 | AQUAfast total nitrogen HR digestion tubes, 50 pack | 5 to 150mg/L N |
ACD095 | AQUAfast phosphate (total) digestion tubes, 50 pack | 0.02 to 1.1mg/L P |
ACD095AH | AQUAfast phosphate (acid hydrolysable) digestion tubes, 50 pack | 0.02 to 1.6mg/L P |
ACR007 | AQUAfast nitrate reaction tubes, 50 pack | 1 to 30mg/L N |
ACR011 | AQUAfast ammonia HR reaction tubes, 50 tubes | 1 to 50mg/L N |
ACR012 | AQUAfast ammonia LR reaction tubes, 50 tubes | 0.02 to 2.5mg/L N |
ACR095 | AQUAfast phosphate (ortho) reaction tubes, 50 pack | 0.06 to 5mg/L PO4 |
CODH00 | AQUAfast COD 0-1500 ppm digestion tubes, 25 pack | 0 to 1500mg/L O2 |
CODHP0 | AQUAfast COD 0-15000 ppm digestion tubes, 25 pack | 0 to 15000mg/L O2 |
CODL00 | AQUAfast COD 0-150 ppm digestion tubes, 25 pack | 0 to 150mg/L O2 |
CODL150 | AQUAfast COD 0-150 ppm digestion tubes, 150 pack | 0 to 150 mg/L O2 |
CODM150 | AQUAfast COD 0-1500 ppm digestion tubes, 150 pack | 0 to 1500 mg/L O2 |