Search Thermo Fisher Scientific
Catalog Number | Bed Length | Bed Width | Flow Rate | Pillar Length |
---|---|---|---|---|
COL-CAP050G1B | 50 cm | 1 mm | 1 to 15 μL/min | 28 μm |
COL-NANO200G1B | 200 cm | 315 μm | 0.1 to 1 μL/min | 18 μm |
Sample Coverage - μPACᵀᴹ HPLC columns provide comprehensive coverage that is ideal for proteomics, metabolomics, and lipidomics. When used for single shot proteome analysis, nearly 10,000 identifications can be achieved.
Column-to-Column Reproducibility - Each column is manufactured using the same lithographic mask, providing consistency and performance from column to column.
Flow Rate Flexibility - μPACᵀᴹ HPLC columns can be operated at moderate LC pump pressures up to 350 bar over a wide range of flow rates (100 – 1,000 nL/min).