Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

ChargeSwitch™ PCR Clean-Up Kit

Catalog number: CS1200010
Have Questions?
Invitrogen™

ChargeSwitch™ PCR Clean-Up Kit

Catalog number: CS1200010
Have Questions?
Catalog Number
CS1200010
Unit Size
960 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
CS1200010960 preps
-
CS12000100 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts (Figure 1). This kit is scalable for use on liquid-handling robots. This kit includes all the necessary reagents for the number of preps listed and requires the use of a magnetic accessory. Please see the Accessories Chapter (Chapter 6) to select the best magnetic accessory for your needs.

• Sample size: 25–50 μL
• High recovery of DNA fragments
• Rapid, one tube, 5 minute protocol

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
High-throughput Compatibility
High-throughput Compatible
Product Line
ChargeSwitch™
Product Type
PCR Clean-Up Kit
Quantity
960 Preps
Run Time
5 min
Shipping Condition
Room Temperature
Target
DNA from Enzymatic Reactions (e.g. PCR)
For Use With (Application)
Next-Generation Sequencing (NGS), Cloning, Standard PCR
Isolation Technology
ChargeSwitch™ Chemistry
No. of Reactions
960 Preps
Sample Type
PCR Products

Contents & Storage

The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly.

Components:
• ChargeSwitch™ Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text