Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Thermo Scientific™
Oxoid™ Novobiocin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Have Questions?
變更視圖buttonViewtableView
產品號碼DescriptionAntibiotic CodeConcentration
CT0038BOxoid Novobiocin Antimicrobial Susceptibility DiscsNV30μg
CT0037BOxoid Novobiocin Antimicrobial Susceptibility DiscsNV5μg
產品號碼 CT0038B
價格 (HKD)
-
申請報價
Description:
Oxoid Novobiocin Antimicrobial Susceptibility Discs
Antibiotic Code:
NV
Concentration:
30μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Novobiocin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
規格
Antibiotic CodeNV
Antimicrobial AgentsNovobiocin
Concentration30μg
DescriptionOxoid Novobiocin Antimicrobial Susceptibility Discs
No. per Pack5 Cartridges/Pk.
Packaging Type50 Discs/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach
Intended use statement
Oxoid Antimicrobial Susceptibility Test Discs are used in the semi-quantitative agar diffusion test method for in vitro susceptibility testing. Used in a diagnostic workflow to aid clinicians in determining potential treatment options for patients suspected of having a microbial infection, these discs are intended to determine susceptibility against microorganisms for which novobioin has been shown to be active both clinically and in vitro. To be used with a pure, agar grown culture, Oxoid Antimicrobial Susceptibility Test Discs are for professional use only and are neither automated, nor a companion diagnostic.
Please enter sku to load default questions
文件與下載
證書
安全資料表
分享目錄編號、產品名稱或連結。