Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks, 40 μg
Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks, 40 μg
Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks, 40 μg
Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks, 40 μg
Thermo Scientific™
Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks, 40 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Have Questions?
Catalog NumberDescriptiondimAntibioticCodeConcentration
CT1755BOxoid Penicillin/Novobiocin Antimicrobial Susceptibility Disks40μg
Catalog number CT1755B
Price (USD)
-
Request A Quote
Description:
Oxoid Penicillin/Novobiocin Antimicrobial Susceptibility Disks
Concentration:
40μg
Ask our AI about this Product
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Penicillin/Novobiocin Antimicrobial Susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For Veterinary Use Only.
For Veterinary Use Only.
Specifications
Antibiotic CodePNV
Antimicrobial AgentsPenicillin/Novobiocin
Certifications/ComplianceComplies with CLSI guidelines
Concentration40μg
DescriptionOxoid Penicillin/Novobiocin Antimicrobial Susceptibility Disks
No. per Pack5 Cartridges/Pk.
Packaging Type50 Disks/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link