IL2 (Interleukin 2) Recombinant Human Protein
Product Image
Gibco™

IL2 (Interleukin 2) Recombinant Human Protein

Recombinant Human Interleukin 2 (IL2) is a bioactive protein intended for use in cell culture applications. IL2 causes proliferation ofRead more
Have Questions?
Catalog number CTP0021
Price (EUR)
1 794,00
100 µg
-
Add to cart
Price (EUR)
1 794,00
100 µg
Add to cart
Ask our AI about this Product
Recombinant Human Interleukin 2 (IL2) is a bioactive protein intended for use in cell culture applications. IL2 causes proliferation of T cells and is a central regulator of immune responses. The lyophilized, E. coli-expressed IL2 Recombinant Human is verified for activity through dose-dependent proliferation of mouse CTLL-2 cells.

Highly Pure—minimizes interference from other proteins or contaminants
Active—get more results with less protein

High Purity
To help verify that GIBCO™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ recombinant growth factor is verified by SDS-PAGE to be over 95% pure and to have endotoxin levels below 0.1 ng/μg.

High Biological Activity
All GIBCO™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our IL2 Recombinant Human growth factor is determined by the dose dependent proliferation of mouse CTLL-2 cells to have an ED50 range of 0.08–0.5 ng/mL. Optimal concentration for individual applications should be determined by a dose response assay.

Related Links
Our Full Collection of GIBCO™ Growth Factors
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Cell Culture
Molecular Weight (g/mol)15.5 kDa
Purification MethodGel-Purified
Quantity100 μg
SourceHuman
Gene AliasIL-2 (Interleukin 2)
SpeciesHuman
RecombinantRecombinant
ClassificationCarrier-Free
Activity0.281 ng/ml
BioassayProliferation of CTLL-2 cells
Endotoxin Level< 0.1 ng/μg
ReactivityHuman
Shipping ConditionWet Ice
Expression SystemE. coli
Protein FamilyCytokines & Receptors
Protein FormRecombinant, Ligand, Mutant
Protein SubtypeInterleukins
Research CategoryClinical Research, Signal Transduction, Oncology, Stem Cell Research, Drug Development, Immunology, Angiogenesis
Product LineGibco™
Purity or Quality Grade95 %
Unit Size100 µg
Contents & Storage
Lyophilized protein can be stored at 2-8°C dessicated. Once reconstituted, store in aliquots at ≤20°C. Avoid multiple freeze⁄thaw cycles.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection