Specifications
Activity0.855 ng/ml
BioassayProliferation of human PBMCs
ClassificationCarrier-Free
Endotoxin Level< 0.1 ng/μg
Expression SystemE. coli
Gene AliasIL-7 (Interleukin 7)
Molecular Weight (g/mol)17.5 kDa
Product LineGibco™
Protein FamilyCytokines & Receptors
Protein FormFull Length, Recombinant, Ligand
Protein SubtypeInterleukins
Purification MethodGel-Purified
Purity or Quality Grade95 %
Quantity100 μg
ReactivityHuman
Research CategoryClinical Research, Signal Transduction, Stem Cell Research, Drug Development, Immunology, Oncology
Shipping ConditionWet Ice
For Use With (Application)Cell Culture
RecombinantRecombinant
SpeciesHuman
Unit Size100 µg