Gibco™

CTS™ Vitronectin (VTN-N) Recombinant Human Protein, truncated

Catalog number: CTS279S3
Have Questions?
Gibco™

CTS™ Vitronectin (VTN-N) Recombinant Human Protein, truncated

Catalog number: CTS279S3
Have Questions?
Catalog Number
CTS279S3
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
CTS279S31 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco CTS (Cell Therapy Systems) Vitronectin (VTN-N) Recombinant Human Protein provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs). When used with CTS Essential 8 Medium (Cat. No. A2656101), CTS Vitronectin has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines. In addition, CTS Vitronectin has been shown to support PSC growth for >8 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages. The VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8 Medium (1).

Features of CTS Vitronectin include:
Designed for clinical research—USP Chapter 1043* and Ph Eur 5.2.12** considered in the design
Animal origin–free—for reduced variability and contamination risk in PSC cultures
Optimized—designed for use with CTS Essential 8 Medium
Consistent—supports seamless transition from RUO Vitronectin
Traceable—documentation includes COO and COA
Controlled—manufactured in compliance with ISO-13485

Designed for cell and gene therapy development and manufacturing
Both USP Chapter 1043 and Ph. Eur. 5.2.12 have been considered in the design of CTS Vitronectin, and this product is the only such product labeled with 'For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration to humans or animals.' Traceability documentation is provided with the product and includes a Certificate of Analysis (COA) and Certificate of Origin (COO) to support regulatory filings.

Animal origin–free system
CTS Vitronectin is animal origin–free to help reduce contamination risk and variability associated with animal-origin raw materials compared to human plasma–derived vitronectin and standard basement membrane extracts (BMEs).

Optimized and defined surface
Unlike standard BMEs, CTS Vitronectin is based on the N-variant design of vitronectin (VTN-N) from the laboratory of James Thomson for use with Essential 8 Medium and has been shown to support human pluripotent stem cell attachment and survival better than wild type vitronectin. CTS Vitronectin provides an optimal environment for feeder-free pluripotent stem cell culture.

Seamless transition from bench to clinic
PSC cultures expanded in other feeder or feeder-free systems can be transitioned to CTS Essential 8 Medium and CTS Vitronectin with minimal adaptation.

Manufacturing control and traceability
CTS Vitronectin is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified. Full traceability documentation such as Certificate of Origin and Certificate of Analysis are available.

Commercialized in partnership with Cellular Dynamics International.

1. Chen G, Gulbranson DR, Hou Z, Bolin JM, Ruotti V, Probasco MD, Smuga-Otto K, Howden SE, Diol NR, Propson NE, Wagner R, Lee GO, Antosiewicz-Bourget J, Teng JM, Thomson JA. Chemically defined conditions for human iPSC derivation and culture. Nat Methods. 2011 May;8(5):424-9.

* U. S. Pharmacopeia. Chapter 1043: Ancillary materials for cell, gene, and tissue-engineered products.
** European Pharmacopeia. Chapter 5.2.12: Raw materials of biological origin for the production of cell-based and gene therapy medicinal products.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Specifications

Concentration
0.81–0.99 mg/mL
For Use With (Application)
Cell and Gene Therapy Research, Development, and Manufacturing
Shelf Life
36 months from date of manufacture
Product Type
Pluripotent Stem Cell Matrix
Species
Human
Sterility
Sterile-filtered
Classification
Animal Origin-Free
Shipping Condition
Dry Ice
Expression System
E. coli
Manufacturing Quality
ISO 13485

Contents & Storage

1 mL cryopreserved vial

Store at -70°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text