The bright green fluorescent Alexa Fluor™ 488 DNase I selectively binds to G-actin (globular) and can be used for the detection of unpolymerized actin in fixed cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity
5 mg
Conjugate
Alexa Fluor 488
Excitation/Emission
495⁄519
Label or Dye
Alexa Fluor™ 488
Label Type
Alexa Fluor Dyes
Shipping Condition
Room Temperature
Sub Cellular Localization
Actin, Cytoskeleton
Form
Lyophilized
Product Line
Alexa Fluor™
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.