Invitrogen™

DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

Catalog number: D2248
Have Questions?
Invitrogen™

DNP-X, SE, 6-(2,4-Dinitrophenyl)aminohexanoic Acid, Succinimidyl Ester

Catalog number: D2248
Have Questions?
Catalog Number
D2248
also known as D-2248
Unit Size
25 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D2248
also known as D-2248
25 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The amine-reactive DNP-X succinimidyl ester can be used bioconjugates that can be detected with anti-dinitrophenyl (anti-DNP) antibodies. This reactive hapten contains an additional seven-atom aminohexanoyl spacer ('X') between the fluorophore and the succinimidyl ester group. This spacer helps to separate increase accessibility to anti-DNP antibodies.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
25 mg
Product Type
DNP-X Succinimidyl Ester
Chemical Reactivity
Amine
Shipping Condition
Room Temperature
Reactive Moiety
Active Ester, Succinimidyl Ester
Label or Dye
DNP
Label Type
Biotin and Other Haptens

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text