Dextran, Alexa Fluor™ 647; 10,000 MW, Anionic, Fixable
Dextran, Alexa Fluor™ 647; 10,000 MW, Anionic, Fixable
Invitrogen™

Dextran, Alexa Fluor™ 647; 10,000 MW, Anionic, Fixable

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D229142 mg
Catalog number D22914
Price (EUR)
391,65
Online Exclusive
424,00
Save 32,35 (8%)
Each
-
Add to cart
Quantity:
2 mg
Recurring order eligible. Learn more »
Price (EUR)
391,65
Online Exclusive
424,00
Save 32,35 (8%)
Each
Add to cart
Ask our AI about this Product
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

Label (Ex/Em): Alexa Fluor™ 647 (650/668)
Size: 10,000 MW
Charge: Anionic
Fixable: Fixable via free amines

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates-Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor, Cascade Blue, lucifer yellow, fluorescein, and Oregon Green dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

Neuronal tracing (anterograde and retrograde) in live cells
Cell lineage tracing in live cells
Neuroanatomical tracing
Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
Investigating vascular permeability and blood-brain barrier integrity
Tracking endocytosis
Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeAlexa Fluor Dyes
Product TypeDextran
Quantity2 mg
Shipping ConditionRoom Temperature
Excitation/Emission650/668 nm
Product LineAlexa Fluor™
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
What are the Gauss levels for the DynaMag-2 and DynaMag-Spin Magnets?
What is the difference between the DynaMag-2 Magnet (Cat. No. 12321D) and DynaMag-Spin Magnet (Cat. No. 12320D)?
What is the standard conversion of Gauss to Tesla units?
How can I clean my DynaMag magnet?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Fluorescence spectra

Fluorescence spectra

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3011756Certificate of AnalysisOct 17, 2024D22914
2860812Certificate of AnalysisFeb 02, 2024D22914
2559189Certificate of AnalysisMar 02, 2023D22914
2403707Certificate of AnalysisNov 10, 2021D22914
2291623Certificate of AnalysisApr 12, 2021D22914
5 results displayed, search above for a specific certificate

Safety Data Sheets

Product Information

Frequently asked questions (FAQs)

If you want to see the most detailed structure you should use the low molecular weight conjugated dextrans such as the 3,000 MW dextrans.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Ensure that the dextran you are using is the fixable form (i.e., contains a primary amine). Dextrans that do not contain a primary amine will not be fixed. Another factor could be that the concentration of the dextran is too low, and the concentration use can be increased up to 10 mg/mL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

We do not determine the net charge of the dextran conjugates. The net charge depends on the fluorophore used to label the dextran and the method of preparing the conjugate. We label some dextrans as neutral or anionic based on the fluorophore used, however the net charge of the dextran may not always be the same as the dye. The Alexa Fluor, Cascade Blue, Lucifer Yellow, fluorescein, and Oregon Green dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic Rhodamine B, tetramethylrhodamine and Texas Red dyes are essentially neutral.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Dextrans with molecular weights from 3,000 to 70,000 have been used, however the 3,000 and 10,000 MW dextrans are most commonly used for neuronal tracing. The 3,000 MW dextrans are used for more detailed tracing of fine neuronal projections, investigating gap junctions, and diffuse more quickly; while the 10,000 MW dextrans have slower distribution, longer cellular retention, and do not cross gap junctions.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

The NeuroTrace BDA-10,000 Neuronal Tracer Kit (Cat. No. N7167) manual has a good protocol for injection procedures and neuronal tracing using the10,000 MW lysine-fixable biotin dextran amine (BDA). This protocol could potentially be applied to other fluorescent dextrans.

Please review Tables 1a and 1b on pages 4 and 5 - https://tools.thermofisher.com/content/sfs/manuals/mp07167.pdf

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (33)

Citations & References
Abstract
Glycosylation influences the lectin activities of the macrophage mannose receptor.
Authors:Su Y, Bakker T, Harris J, Tsang C, Brown GD, Wormald MR, Gordon S, Dwek RA, Rudd PM, Martinez-Pomares L
Journal:J Biol Chem
PubMed ID:15983039
'The mannose receptor (MR) is a heavily glycosylated endocytic receptor that recognizes both mannosylated and sulfated ligands through its C-type lectin domains and cysteine-rich (CR) domain, respectively. Differential binding properties have been described for MR isolated from different sources, and we hypothesized that this could be due to altered glycosylation. ... More
Receptor (CD155)-dependent endocytosis of poliovirus and retrograde axonal transport of the endosome.
Authors:Ohka S, Matsuda N, Tohyama K, Oda T, Morikawa M, Kuge S, Nomoto A
Journal:J Virol
PubMed ID:15194795
'Poliovirus (PV), when injected intramuscularly into the calf, is incorporated into the sciatic nerve and causes an initial paralysis of the inoculated limb in transgenic mice carrying the human PV receptor (hPVR/CD155) gene. Here, we demonstrated by using an immunoelectron microscope that PV particles exist on vesicle structures in nerve ... More
Characterizing peptide-mediated DNA internalization in human cancer cells.
Authors:Wittrup A, Belting M,
Journal:Methods Mol Biol
PubMed ID:19085116
'Cell penetrating peptides (CPPs) are currently used to deliver various macromolecular cargos to intracellular sites of action both in vitro and in vivo on an experimental basis. During the last few years, even more evidence has accumulated indicating that the main route of entry for most CPPs is through endocytosis ... More
Control of cellular physiology by TM9 proteins in yeast and Dictyostelium.
Authors:Froquet R, Cherix N, Birke R, Benghezal M, Cameroni E, Letourneur F, Mösch HU, De Virgilio C, Cosson P,
Journal:J Biol Chem
PubMed ID:18178563
'TM9 proteins constitute a well defined family, characterized by the presence of a large variable extracellular domain and nine putative transmembrane domains. This family is highly conserved throughout evolution and comprises three members in Dictyostelium discoideum and Saccharomyces cerevisiae and four in humans and mice. In Dictyostelium, previous analysis demonstrated ... More
A stepwise dissection of the intracellular fate of cationic cell-penetrating peptides.
Authors:Fischer R, Köhler K, Fotin-Mleczek M, Brock R
Journal:J Biol Chem
PubMed ID:14707144
'The role of endosomal acidification and retrograde transport for the uptake of the highly basic cell-penetrating peptides penetratin, Tat, and oligoarginine was investigated. The effect of a panel of drugs that interfere with discrete steps of endocytosis or Golgi-mediated transport on uptake and cellular distribution of fluorescein-labeled peptide analogues was ... More
33 total citations

Other products to consider



Share catalog number, name or link