PED6 (N-((6-(2,4-Dinitrophenyl)amino)hexanoyl)-2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Product Image

PED6 (N-((6-(2,4-Dinitrophenyl)amino)hexanoyl)-2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

Have Questions?
Catalog number D23739
also known as D-23739
Price (USD)/ 1 mg
2,100.00
-
Add to cart
Price (USD)/ 1 mg
2,100.00
Add to cart
PED6 (N-((6-(2,4-Dinitrophenyl)amino)hexanoyl)-2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Catalog numberD23739
Price (USD)/ 1 mg
2,100.00
-
Add to cart
Molecular Probes® phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells. Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.

Fluorogenic Phospholipase A Substrates Specifications:
• Label (Ex/Em): PED6
• Specificity: PLA2 (this substrate is cleaved by PLA1, but this reaction does not separate the fluorophore from the quencher)
• Cleavage product: D3834
• Lyophilized product can be dissolved in DMSO for use
• Fluorescence is measured either in microplate assays or using flow cytometry


Find More Probes for Lipid Metabolism and Fatty Acid Analogs
Review Fatty Acid Analogs and Phospholipids—Section 13.2 and Probes for Lipid Metabolism and Signaling—Section 17.4 in the Molecular Probes® Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Size1 mg
Reagent TypePhospholipids
Shipping ConditionRoom Temperature
Unit Size1 mg