Invitrogen™

DAF-FM (4-Amino-5-Methylamino-2',7'-Difluorofluorescein)

Catalog number: D23841
Have Questions?
Invitrogen™

DAF-FM (4-Amino-5-Methylamino-2',7'-Difluorofluorescein)

Catalog number: D23841
Have Questions?
Catalog Number
D23841
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D238411 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
DAF-FM is a reagent that is used to detect and quantify low concentrations of nitric oxide (NO). It is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. DAF-FM fluorescence can be detected by any instrument that can detect fluorescein, including flow cytometers, microscopes, fluorescent microplate readers and fluorometers.

DAF-FM Specifications:
• Ex/Em of DAF-FM: ∼495/515 nm
• Lyophilized product should be dissolved using DMSO and then added to an aqueous buffer to create a working solution
• DAF-FM can be introduced into cells by pressure injection or perfusion from a patch-clamp pipette (choose DAF-FM diacetate if you need a cell-permeant version)
• Buffers containing bovine serum albumin (BSA) and phenol red may affect the fluorescence and should be used with caution
• The fluorescence quantum yield of DAF-FM is ∼0.005, but increases about 160-fold, to ∼0.81, after reacting with NO


Applications of DAF-FM:
• Assessment of NO production in transaldolase-deficient lymphoblasts by flow cytometry
• Detection of NO accumulation in embryonic cortical neurons following neurotrophin stimulation
in vivo imaging of NO in zebrafish
• Intravital microscopic detection of NO generation associated with angiogenesis in mice
• Quantitation of ATP-induced NO release in rabbit platelets

Important Advantages of DAF-FM Over DAF-2
The spectra of the NO adduct of DAF-FM are independent of pH above pH 5.5. Also, the NO adduct of DAF-FM is significantly more photostable than that of DAF-2, which means additional time for image capture. Finally, DAF-FM is a more sensitive reagent for NO than is DAF-2 (NO detection limit for DAF-FM ∼3 nM versus ∼5 nM for DAF-2).

Probes for Nitric Oxide Research
We offer several products for the detection of reactive oxygen species (ROS) and nitric oxide (NO). Review these products in Probes for Nitric Oxide Research—Section 18.3 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Properties
Fluorescent Dye-Based
Shipping Condition
Room Temperature
Excitation/Emission
495/515 nm
Indicator
Free Radical Sensor
Product Line
Invitrogen™

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text