Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Catalog Number | Modules | Voltage |
---|---|---|
D4631 | 3 | 120 V |
D4632-33 | 3 | 240 V |
D4641 | 4 | 120 V |
D4642-33 | 4 | 240 V |
Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.
AquaPro™ digital on-line analytical measurement platform featuring:
Ordering Information:
Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.
Includes:
Wall bracket
Requires:
O-rings included with cartridge pack (sold separately)
Recommended for:
AA, GS, buffer preps, feedwater for analytical instruments or labware washers
Note:
If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).
Discover how our water purification systems deliver pure and ultrapure water - with ease.
Let our water experts recommend the ideal system for your needs with an H2O Select water analysis.
Share catalog number, name or link