Search Thermo Fisher Scientific
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
Description | Cartridge pack; deionized feed |
Feedwater Source | Deionized |
For Use With (Equipment) | Easypure™ II and Easypure UF |
Type | Cartridge Pack |
Unit Size | Each |
Description | Cartridge pack; deionized feed |
Feedwater Source | Deionized |
For Use With (Equipment) | Easypure™ II UV and Easypure II UV/UF |
Type | Cartridge Pack |
Unit Size | Each |
Description | Cartridge pack; RO or distilled feed |
Feedwater Source | Reverse Osmosis, Distilled |
For Use With (Equipment) | Easypure™ II UV and Easypure II UV/UF |
Type | Cartridge Pack |
Unit Size | Each |
Description | Cartridge pack; RO or distilled feed |
Feedwater Source | Reverse Osmosis, Distilled |
For Use With (Equipment) | Easypure™ II and Easypure UF |
Type | Cartridge Pack |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Description | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
D502126 | Each | Easypure™ II and Easypure UF | Cartridge pack; deionized feed | 636.30 Online price 707.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D502124 | Each | Easypure™ II UV and Easypure II UV/UF | Cartridge pack; deionized feed | 655.20 Online price 728.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D502125 | Each | Easypure™ II UV and Easypure II UV/UF | Cartridge pack; RO or distilled feed | 655.20 Online price 728.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D502127 | Each | Easypure™ II and Easypure UF | Cartridge pack; RO or distilled feed | 599.40 Online price 666.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
D502126:
D502124:
D502127:
D502125: