Invitrogen™

Di-2-ANEPEQ (JPW 1114)

Catalog number: D6923
Have Questions?
Invitrogen™

Di-2-ANEPEQ (JPW 1114)

Catalog number: D6923
Have Questions?
Catalog Number
D6923
also known as D-6923
Unit Size
5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
D6923
also known as D-6923
5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ANEP dyes are molecules that fluoresce in response to electrical potential changes in their environment. These are fast-response probes that operate by means of a change in their electronic structure, and consequently their fluorescence properties, in response to a change in the surrounding electric field. Their optical response is sufficiently fast to detect transient (millisecond) potential changes in excitable cells, including single neurons, cardiac cells, and intact brains. However, the magnitude of their potential-dependent fluorescence change is often small; fast-response probes typically show a 2–10% fluorescence change per 100 mV. Furthermore, these dyes display a potential-dependent shift in their excitation spectra, thus permitting the quantitation of membrane potential using excitation ratio measurements

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›

Potential-Sensitive ANEP Dye Specifications:
• Ex/Em maxima bound to model phospholipid membranes are ∼465/635 nm (but spectral properties are highly dependent on environment)
• Nonfluorescent until bound to membranes
• Cationic molecule; soluble in water (di-2-ANEPEQ is a water-soluble ANEP dye)
• Dye is typically introduced into cells by microinjection
• Fast-response probe, suitable for detecting submillisecond membrane potential changes


Applications for Potentiometric Probes
The plasma membrane of a cell typically has a transmembrane potential of approximately –70 mV (negative inside) as a consequence of K+, Na+, and Cl– concentration gradients that are maintained by active transport processes. Potentiometric probes offer an indirect method of detecting the translocation of these ions.

Increases and decreases in membrane potential—referred to as membrane hyperpolarization and depolarization, respectively—play a central role in many physiological processes, including nerve-impulse propagation, muscle contraction, cell signaling and ion-channel gating. Potentiometric probes are important tools for studying these processes.

Find More ANEP Dyes
We offer ANEP dyes in various forms. Review Fast-Response Probes—Section 22.2 in the Molecular Probes™ Handbook for more information on these probes.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Infrared
Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope, Microplate Reader
Product Type
ANEP Dye
Quantity
5 mg
Shipping Condition
Room Temperature
Sub Cellular Localization
Cell Membranes & Lipids

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text