Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product Type | Printer |
For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Dispensing Platform |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Dispensing Platform |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Trocar |
For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Bag Stand |
Description | Stand for broth bag |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Bag Stand |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Tube Holding Arm |
Description | Mobile tube holding arm |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Tube Holding Arm |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Bottle |
Capacity (Metric) | 5 L |
Description | 5 L bottle (autoclavable) |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Bottle |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Calibration Weight |
Capacity (Metric) | 500 g |
Description | 500 g calibration weight |
For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Foot Pedal |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Foot Pedal |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Nozzle |
Description | Stainless steel nozzle (5 pieces) |
For Use With (Equipment) | Diluflux Diluter, Homogenizer, Nozzle |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Product Type | Weight |
Description | Tube weight (5 pieces) |
For Use With (Equipment) | Diluflux™ Diluter and Homogenizer |
Packaging Quantity | 1 Ea. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Product Type | Packaging Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
DB4008A | Each | Printer | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4001A | Each | Dispensing Platform | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4002A | Each | Trocar | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4003A | Each | Bag Stand | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4004A | Each | Tube Holding Arm | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4005A | Each | Bottle | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4006A | Each | Calibration Weight | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4007A | Each | Foot Pedal | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4009A | Each | Nozzle | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
DB4010A | Each | Weight | 1 Ea. | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|