Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Product Type | Packaging Quantity |
---|---|---|
DB4017A | Printer Roll | 2 Rolls |
DB4001A | Dispensing Platform | 1 Ea. |
DB4002A | Trocar | 1 Ea. |
DB4003A | Bag Stand | 1 Ea. |
DB4004A | Tube Holding Arm | 1 Ea. |
DB4005A | Bottle | 1 Ea. |
DB4006A | Calibration Weight | 1 Ea. |
DB4007A | Foot Pedal | 1 Ea. |
DB4008A | Printer | 1 Ea. |
DB4009A | Nozzles | 5 pieces/Bag |
DB4010A | Weight | 1 Ea. |
DB4016A | Printer Stickers | 2 Rolls, 980/Roll |
DB4018A | Dispensing Assembly | 1 Ea. |
DB4019A | Tubing Set | 1 Ea. |
DB4020A | Pump | 1 unit |
DB5001A | Bag Stand | 1 Ea. |
DB5002A | Bag Stand | 1 rack |
DB5003A | Bag Clips | 50 clips |