Search Thermo Fisher Scientific
Catalog Number | Concentration | Packaging Type | Quantity |
---|---|---|---|
DD0001B | 5µg | 5 Cartridges/Pk. | 50 Discs/Cartridge |
Thermo Scientific™ Oxoid Optochin Disc is for the differentiation of Streptococcus pneumoniae.
Differentiate Streptococcus pneumoniae from other streptococci using Thermo Scientific™ Oxoid™ Optochin Disc. Optochin Discs are paper discs ready impregnated with Optochin (ethylhydrocuprein hydrochloride) which provide a convenient and reliable alternative to the bile-solubility test. Pneumococciare are sensitive to Optochin so that a culture shows a zone of inhibition around the impregnated disc, while streptococci either grow right up to the edge of the disc or, occasionally, show a very small zone of inhibition.
Ordering Alerts:
5μg. 50 Discs/Cartridge. 1 Cart/Pk.
Impregnated with 'Optochin' (ethylhydrocuprein hydrochloride) and used in a diagnostic workflow to aid Microbiology professionals in identifying potential pathogenic bacteria suspected in patients having a Streptococcus pneumoniae infection. These discs provide a qualitative result that enables the differentiation of Streptococcus pneumoniae from other alpha-haemolytic streptococci. To be used with a pure, agar grown culture, the device is for professional use only and is neither automated, nor a companion diagnostic.
Share catalog number, name or link