Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
DD0004T | 50 Disks/Cartridge |
Thermo Scientific™ Oxoid V Factor Disc is used for identification of Haemophilus spp.
Ordering Alerts:
50 Discs/Cartridge
Impregnated with X and/or V factor and used in a diagnostic workflow to aid Microbiology professionals in identifying potential pathogenic bacteria suspected in patients having a Haemophilus species infection. These discs enable to growth of Haemophilus species and provide a qualitative result. To be used with a pure, agar grown culture, the device is for professional use only and is neither automated, nor a companion diagnostic.
Share catalog number, name or link