Thermo Scientific™

DynaDrive™ Single-Use Bioreactor, 120 V

Catalog number: DDB0050.1011
Have Questions?
Thermo Scientific™

DynaDrive™ Single-Use Bioreactor, 120 V

Catalog number: DDB0050.1011
Have Questions?
Catalog Number
DDB0050.1011
Unit Size
50 L
Price (USD)
Catalog NumberUnit SizePrice (USD)
DDB0050.101150 LRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific DynaDrive Single-Use Bioreactor (S.U.B.) drives bioprocessing efficiency and allows users to reimagine bioreactor possibilities. The DynaDrive S.U.B. utilizes a revolutionary stir-tank design to meet cell culture demands of today and the future. The DynaDrive S.U.B. not only offers the advantages of single use, but also provides the mixing and mass transfer performance that demanding cell lines require. The hardware, mixing impeller (drive train), and sparging approach have been redesigned to optimize mixing dynamics, scale, and performance. The DynaDrive technology allows for lower seed-train turndown ratios, which facilitates efficiencies and reduces risk of contamination. The complete line of DynaDrive S.U.B.s include 50, 500, 3000, and 5000 L sizes that ensure consistent scalability from pilot-scale studies to pre-clinical to commercial production. The DynaDrive technology brings single-use to unprecedented volumes and performance.

Features:
• Hardware is jacketed with load cells
• Large volume—50 L, 500 L, and option of scalability to larger sizes of up to 5,000 L
• Improved seed train process—10:1 turndown ratio in the 50 L reactor and 20:1 turndown in the 500 L, plus projected larger-size reactors allow for streamlined seed-train process and help reduce risk
• Mixing times, power input per volume (PIV), and kLa performance are optimized for modern cell culture processes with >100 million cells/mL
• Hardware optimized for perfusion cell culture processes
• Ergonomically friendly hardware—designed with the user in mind and allows for consistent and uniform bioprocess container (BPC) loading
• Open-architecture approach—allows interoperability with available control systems
• DynaDrive BPCs with integrated drive train and made with highly robust Thermo Scientific Aegis 5-14 bioprocessing film
• BPC film and components subjected to leachable and extractable (L&E) evaluation according to BioPhorum Operations Group (BPOG) guidelines

Applications of the Thermo Scientific DynaDrive S.U.B. include:
• Batch, fed batch, and perfusion cultures
WARNING: Cancer – www.P65Warnings.ca.gov

Specifications

Capacity (English)
13.2 gal.
Capacity (Metric)
50 L
Height (English)
7 ft. 8 in.
Height (Metric)
2 m 32 cm
Length (English)
2 ft. 7 in.
Material
Stainless Steel
Length (Metric)
77 cm
Model
DynaDrive
Width (English)
2 ft. 4 in.
Width (Metric)
69 cm
Disposable
Reusable

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text