Defender™ Omega Handheld Raman Analyzer
Defender™ Omega Handheld Raman Analyzer
Thermo Scientific™

Defender™ Omega Handheld Raman Analyzer

Perform safe, fast identification of explosives, narcotics and other chemicals with this handheld Raman analyzer.
Have Questions?
Catalog number DEFENDEROMEGA
Price (USD)/ Each
-
Request A Quote
Perform safe, fast identification of explosives, chemical warfare agents, narcotics, and other chemicals with the Thermo Scientific Defender™ Omega Handheld Raman Analyzer. This truly flexible handheld analyzer offers user configurable scan profiles and expandable libraries.

Customs, military and law enforcement personnel can’t always predict what they’ll come across in the field. The Defender Omega analyzer is a reliable handheld tool that can help users identify compounds used to make explosives, nerve agents, narcotics, and other chemicals. This analyzer has all the key elements for essential chemical ID where security and safety professionals need it most:


User safety

  • Touchless, point and shoot scanning through transparent glass or plastic reduces the need for users to handle unknown substances
  • Instructive results deliver an alarm, warning or clear indication to help users quickly determine what action to take

Accuracy

  • Patented chemometric algorithms are more reliable for chemical identification, particularly for multicomponent mixtures and spectrally complex samples
  • Background fluorescence is suppressed to help users confidently identify substances from clear spectral fingerprints using 1064 nm optics

User flexibility

  • The large touchscreen and illuminated keypad allow users to easily enter information, even in low-light situations
  • 2 different scan modes (ID or Screener) enable users of all levels to make fast decisions
  • Flexibility to support user-created libraries to adapt to evolving threats

Comprehensive libraries

  • Expandable libraries help identify explosives, chemical warfare agents, controlled substances, cutting agents, precursors and industrial chemicals
  • Customizable libraries allow users to identify targeted materials
  • Ongoing library updates help users stay current with emerging threats

Solid chain of custody

  • GPS and digital camera enable time and location stamping 
  • Unique user ID and password ensures data stays locked and safe
  • User-generated metadata for fast, easy search of previous measurements
  • Instrument logs to keep track of who scanned what substances and when

Built for use in the field 

  • Removable and rechargeable batteries for power on the go
  • Wi-Fi connectivity for data sharing
  • Web user interface (WebUI) for easy configuration 
  • Watertight – (IP68 rated) for protection against water and dust 
  • MIL-STD-81H rated for durability

Service Plans

Thermo Fisher Scientific offers service plans designed to maximize productivity and optimize performance throughout the lifecycle of your Defender Omega analyzer. Please speak with your sales representative to learn more about service and support options. 


Plans include exclusive service advantages such as:

  • Convenience: unlimited technical support and remote diagnostics
  • Cost control: budget predictability and avoidance of unexpected repair costs
  • Quality repairs: depot and parts availability, with timely turnaround
  • Rapid response: access to around-the-clock, reachback support
  • Reliability: software and performance verifications
Specifications
Battery TypeRemoveable and Lithium Ion, 7.2V, 49Wh, 4-hr run-time at 25°C
Certifications/ComplianceMIL-STD-810H, IP68 rated
ConnectionsWi-Fi and USB-C cable
Dimensions (L x W x H)24.1 x 11.7 x 5.6 cm (9.5 x 4.6 x 2.2 in.)
Includes5 MP digital camera, global positioning, WebUI
IP RatingIP68
Laser Excitation Wavelength1064.25 nm ±0.5 nm, 2 cm-1
Laser OutputAdjustable 480 mW, 240 mW, 96 mW
Ocular Hazard Distance160 cm (63 in.)
Temperature Range (Metric)-20°C to 50°C
Weight (English)3.5 lb.
Weight (Metric)1.6 kg
Unit SizeEach